ContactUsLinkComponent
Glass Conical Philips Beaker with Spout
Glass Conical Philips Beaker with Spout
Catalog # 470344-936
Supplier:  Wards
Supplier #: CH0128A
undefined
Glass Conical Philips Beaker with Spout
Catalog # 470344-936
Supplier:  Wards
Supplier Number:  CH0128B
Supplier #: CH0128A
Log in for availability
Select an Option
2.6"
Select an Option
250 ml
Select an Option
Conical Phillips Beaker with Spout
Select an Option
4.2"
Select an Option
470344-936
Select an Option
0.3 lbs
About this item
Unique conical Philips pattern beaker with pouring spouts for viscous solutions, acids, bases, or any other liquid storage needed.
  • High quality borosilicate 3.3 glass
  • Includes spout for easy pouring
  • Enhanced resistance to thermal shock
  • Autoclavable and top rack dishwasher safe
Eisco Labs Borosilicate glass has the following heat tolerances: Strain Point −515 °C | Annealing Point 565 °C | Softening Point 820 °C.
Eisco Labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.