Bovine Angiotensin I Converting Enzyme (ACE) ELISA Kit
Catalog # 77536-610
Supplier: Bioss
New Product
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:
- Primary antibody reactivity:Bovine
- Target protein:Angiotensin I Converting Enzyme (ACE)
- Size:1 kit
- Environmentally Preferable:
- Detection Method:Colorimetric microplate reader primary wavelength 450 nm
- Shelf Life:12 Months
- Detection Range:6.25 - 400 ng/ml
- Storage Temperature:TMB substrate, wash buffer (30X concentrate), and the stop solution should be stored at 4 °C upon receipt, while the other items should be stored at –20 °C.
- Sensitivity:2.15 ng/ml
- Cat. No.:77536-610
- No. of tests:48
Specifications
About this item
- High sensitivity with a broad assay range
- Flexible inputs: Serum, plasma, cell culture supernatant
- Reproducible and specific results