Methyltransferase Dilution Buffers IV
Supplier: Sino Biological
New Product
Certificates
About this item
Solution of buffer used for diluting enzyme protein methyltransferases.
Storage: For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
Specifications
- Storage Temperature:Store product at –20 °C
- Formulation:Frozen solution of buffer containing: 20 mM Tris-HCl, pH 8.0, 50 mM NaCl, 1 mM EDTA, 3 mM MgCl₂,and 0.1 mg/ml BSA