Order Entry
Puerto Rico
ContactUsLinkComponent
 
Methyltransferase Dilution Buffers III
New Product
 
Methyltransferase Dilution Buffers III
New Product

 

  • Storage Temperature:
    Store product at –20 °C
  • Formulation:
    Frozen solution of buffer containing: 50 mM Tris-HCl, pH 7.5, 1 mM EDTA, 5% glycerol and 10 ng/µl BSA