Methyltransferase Dilution Buffers I
Supplier: Sino Biological
New Product
Certificates
About this item
Solution of buffer used for diluting enzyme protein methyltransferases.
Storage: For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
Specifications
- Storage Temperature:Store product at –20 °C
- Formulation:Frozen solution of buffer containing: 50 mM Tris-HCl, pH 9.0 and 10 ng/µl BSA