Brady® PermaSleeve® Low Smoke Zero Halogen Heat-Shrink Labels
Specialized labels are designed for mass transit and marine applications where flame-retardant, low-smoke wire marking materials are required.
- Matte finish
- Flame-retardant, heat-shrinkable, low-smoke, zero-halogen material
- Recommended ribbon: R6000HF
Durable labels are flame-retardant and resistant to temperatures from −30 to +105 °C (−22 to +221 °F). Fast shrinking with a 2:1 shrink ratio; labels permanently adhere to wires quickly with a heat gun.
: RoHS compliant to 2011/65/EU (Pass 2015/863).