Brady® Retro Reflective All-Weather Labels
Versatile labels are ideal for safety applications such as use on personnel, fire suppression equipment, first aid cabinets, or anywhere extra visibility is required.
- Permanent acrylic adhesive
- Gloss finish
- Retro reflective material
- Recommended ribbon: R6000HF
Durable labels are resistant to abrasions, weather, solvents, and extreme temperatures. Pressure-sensitive acrylic adhesive withstands prolonged exposure to tough conditions. Long-lasting labels are able to withstand outdoor conditions for 4 to 6 years on average.