Brady® Aggressive Adhesive Multi-Purpose Nylon Labels
Versatile labels are ideal for labeling tightly-curved surfaces such as wires and cables, as well as general-purpose lab, equipment panels, faceplates, and terminal blocks.
- Permanent acrylic adhesive
- Matte finish
- Autoclavable material; suitable for cryogenic storage
- Halogen-free
- Recommended ribbon: R4300 (94000-380)
Durable labels are designed to withstand extreme temperatures, chemicals, greases, oils, cleaners, and detergents. Permanent adhesive labels stick to smooth, rough, and powder-coated surfaces alike.
: UL recognized to UL969, CSA to C22.2 No. 0.15-95, RoHS compliant to 2011/65/EU (Pass 2015/863).