Brady® FreezerBondz™ Cryogenic Matte Polyester Labels
Supplier: Brady Worldwide
Certificates
About this item
Specialized labels are designed for laboratory use on glass and polypropylene vials, centrifuges, test tubes, straws, and slides.
- Permanent acrylic adhesive
- Matte finish
- Cold-temperature application; suitable for cryogenic storage
- Autoclavable
- Recommended ribbon: R4300 (94000-380)
Durable labels will withstand solvents, smudges, and extreme temperatures, including storage in liquid nitrogen. Made with a permanent acrylic adhesive for use on flat frozen surfaces or wrapping around tubes and vials with a ⅛" overlap.
Certifications: RoHS compliant to 2011/65/EU (Pass 2015/863).
Frequently Bought Together