Orders LinkContactUsLinkComponent
Brady® Solvent-Resistant Polyester Labels
Supplier:   Brady Worldwide
Brady® Solvent-Resistant Polyester Labels
Supplier:  Brady Worldwide
Certificates
Brady® Solvent-Resistant Polyester Labels
Brady® Solvent-Resistant Polyester Labels
2 Options Available Below
Product Details & Documents
Specialized labels are ideal for lab applications where durability to environmental and chemical conditions is key.
  • Permanent acrylic adhesive
  • Matte finish
  • Autoclavable material is suitable for cryogenic storage
  • Recommended ribbon: R4300 (94000-380)
Durable labels are designed to withstand extreme temperatures and short-term exposure to chemicals such as xylene, DMSO, and ethanol. Made with a permanent acrylic adhesive for secure bonding to flat, smooth surfaces such as stainless steel and polypropylene.
 : UL recognised, CSA accepted. Halogen-free (DIN VDE 0472/part 815). Approvals are for white material only.
Product Family Options
Product Information
  • Material
    Color
    For Use With
    Dimensions
    Packaging
    Availability
    Price
  • Polyester
    Black on white
    BMP41;BMP51;BMP53;M511
    0.25H×1.9"W
    160 labels/roll
    Log in for availability
    Log in for availability
    Each
    Each - $65.42
    $65.42
    Each
       $65.42
    Supplier Number: 170774
  • Polyester
    Black on white
    BMP51;BMP53;M511
    0.9H×0.9"W
    280 labels/roll
    Log in for availability
    Log in for availability
    Each
    Each - $99.70
    $99.70
    Each
       $99.70
    Supplier Number: 170887