Brady® QuickFlag™ Labels
Product Details & Documents
Specialized labels are designed with a tapered adhesive side to create easy-to-apply, one-sided wire and cable flags without mismatched edges or exposed adhesive.
- Permanent rubber-based adhesive
- Gloss finish
- Abrasion-resistant, chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, weather-resistant material
- Recommended ribbon: R6000HF
Durable labels are made with a tough polyester material that resists abrasions, temperatures of −40 to +120° C (−40 to +248 °F), and solvents such as fuel, oil, and water. Made with an ultra-aggressive rubber-based adhesive on the tapered side for permanent use.
Certifications: UL recognized to UL969, CSA accepted, RoHS Compliant to 2011/65/EU (Pass 2015/863).
Specifications
- For Use With:BMP41;BMP51;BMP53;M511
- Color:Black on white
- Dimensions:2H×1"W
- Material:Polyester
- Packaging:140 labels/roll
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



