Brady® Water Dissolvable Paper Labels
Supplier: Brady Worldwide
Certificates
About this item
Specialized labels are ideal for laboratories, restaurants, work-in-process product flows, and other temporary identification applications.
- Dissolvable acrylic adhesive
- Matte finish
- Material dissolves in water
- Inkjet print-receptive
- Recommended ribbon: R4300 (94000-380)
Water dissolvable labels are made with a tight-bonding but completely dissolvable acrylic adhesive that dissolves in warm water within 30 seconds, leaving no residue. For material specific performance and properties, please refer to the Technical Data Sheet (TDS).
Certifications: RoHS compliant to 2011/65/EU (Pass 2015/863).