Order Entry
Puerto Rico
ContactUsLinkComponent
 
Kinase Assay Buffers III
New Product
 
Kinase Assay Buffers III
New Product

 

  • Form:
    Liquid
  • Storage Temperature:
    –20 °C
  • Formulation:
    Frozen solution of buffer containing: 200 mM Tris-HCl, pH 7.4, 100 mM MgCl₂ and 0.5 mg/ml BSA. Add to final concentration of 0.25 mM DTT to the aliquot prior to use. Supplied in separate vial.