Order Entry
Puerto Rico
ContactUsLinkComponent
Lipid Dilution Buffers
Lipid Dilution Buffers
Supplier:  Sino Biological
New Product
Certificates
Lipid Dilution Buffers
Supplier:  Sino Biological
New Product

Specifications

  • Storage Temperature:
    Store product at –20 °C
  • Formulation:
    Frozen solution of buffer containing: 25 mM HEPES, pH 7.5, 0.5 mM EGTA, and 0.5% Triton X-100

Specifications

Related Information