Lipid Kinase Buffers
Supplier: Sino Biological
New Product
Certificates
About this item
The 5X stock solution of buffer used for assaying lipid kinase proteins.
Storage: For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
Specifications
- Storage Temperature:Store product at –20 °C
- Formulation:Frozen stock solution of buffer containing: 250 mM HEPES, pH 7.5, 250 mM NaCl, 10 mM MgCl₂, 2.5 mM EGTA, 125 μg/ml BSA and 0.2% Triton X-100. Add to final concentration of 0.25 mM DTT to the aliquot prior to use. Supplied in separate vial.