About this item
STERIS's Septihol® st Alcohol solution provides microbial control and removes residue from hard surfaces. It can be used straight out of the container and be applied poured or sprayed. Since it contains no stabilizers or denaturants, it evaporates quickly and completely.
- Efficacy – Biocidal and fungicidal EPA claims
- Productivity - Ready to use sterile alcohol eliminates in-house production validation and documentation concerns
- Safety - Double bagged and gamma irradiated for ease of introduction into aseptic areas
- Rinsing – Evaporates quickly with no residue
- Reliability - Each lot is tested to ensure sterility and low endotoxins
Septihol® st Alcohol solution is ideal as a final microbial control and disinfection step to remove residue from surfaces and to decontaminate gloves. Its EPA biocidal and fungicidal claims are proven in presence of 5% organic soil on hard, nonporous surfaces. Septihol® st is double-bagged and gamma irradiated for use in aseptic areas. The formula is a blend of 70% (v/v) USP isopropyl alcohol and 30% USP Purified Water. It does not contain stabilizers or denaturants, so it evaporates quickly and completely. Each lot is documented with a Certificate of Analysis for lot traceability.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot Numberghs_6248
SDS
VWR GHS Compliance Letter (US)