ContactUsLinkComponent
Flowflex™ Plus COVID19 Flu A/B Home Kit
Flowflex™ Plus COVID19 Flu A/B Home Kit
Catalog # 77578-758
New Product
CAS Number:  
undefined
Flowflex™ Plus COVID19 Flu A/B Home Kit
Catalog # 77578-758
Supplier Number:  L03A-R0645
New Product
CAS Number:  
Specifications
  • Description:
    Flowflex™ Plus COVID-19 flu A/B home kit
  • Cat. No.:
    77578-758
Specifications
About this item
OTC Covid/Flu combo test kit.
  • Results in 15 minutes
  • 15 months shelf life
  • Small sized packaging makes for easy travel and storage
  • Store between 36 to 86 °F
  • Self-collected anterior nasal swab specimens from individuals aged 14 years or older
  • Sample collection by an adult in children ages 2 to 13
  • Better performance when compared to a FDA EUA and 510K RT-PCR
A rapid test for the detection and differentiation of SARS-CoV-2, Influenza A, and Influenza B protein antigens in anterior nasal swab specimens.