Orders LinkContactUsLinkComponent
TeleSwab Extendable Samplers
TeleSwab Extendable Samplers
TeleSwab Extendable Samplers
Certificates
TeleSwab Extendable Samplers
TeleSwab Extendable Samplers
6 Options Available Below
Product Details & Documents
The TeleSwab has an extendable handle, which allows to take swabs from high and difficult to reach places.
  • Environmental swabbing for use with large items of processing equipment
  • Extendable handle to the correct length for easy swabbing
  • Adapter allowing the swabs to be angled to the contours of the sampled item
Delivery information: Swabs are not included with these kits.
Product Family Options
Product Information
  • Availability
    Price
  • 77576-636
    Telescopic swabbing tool with extendable pole, swab holder and angle adapter
    Telescopic swabbing tool with extendable pole, swab holder and angle adapter
    Log in for availability
    Log in for availability
    Each
    Each - $3,710.00
    $3,710.00
    Each
       $3,710.00
    Supplier Number: 8011K-01
  • 77576-638
    Telescopic swabbing tool with extendable pole, swabbing template, template holder and angle adapter
    Telescopic swabbing tool with extendable pole, swabbing template, template holder and angle adapter
    Log in for availability
    Log in for availability
    Each
    Each - $3,710.00
    $3,710.00
    Each
       $3,710.00
    Supplier Number: 8011K-02
Components
Product Information
  • Availability
    Price
  • 77576-628
    Telescopic pole
    Telescopic pole
    Log in for availability
    Log in for availability
    Each
    Each - $2,030.00
    $2,030.00
    Each
       $2,030.00
    Supplier Number: 8011A-3
  • 77576-630
    Universal swab holder, 316 stainless steel construction
    Universal swab holder, 316 stainless steel construction
    Log in for availability
    Log in for availability
    Each
    Each - $902.00
    $902.00
    Each
       $902.00
    Supplier Number: 8011A-4B
  • 77576-634
    Swabbing template, PTFE, 5×5 cm hole
    Swabbing template, PTFE, 5×5 cm hole
    Log in for availability
    Log in for availability
    Each
    Each - $522.00
    $522.00
    Each
       $522.00
    Supplier Number: 8011A-7-25
  • 77576-632
    Swabbing template, PTFE, 10×10 cm hole
    Swabbing template, PTFE, 10×10 cm hole
    Log in for availability
    Log in for availability
    Each
    Each - $522.00
    $522.00
    Each
       $522.00
    Supplier Number: 8011A-7-100