Orders LinkContactUsLinkComponent
MultiThief Samplers
MultiThief Samplers
Certificates
MultiThief Samplers
MultiThief Samplers
8 Options Available Below
Product Details & Documents
The MultiThief is designed for the multi-level sampling of free flowing powders and granules.
  • Variable sample volumes up to 60 ml
  • Suitable for free fl owing powders and granules
  • Can be quickly and easily changed by removing cells
Product Family Options
Product Information
  • Material
    Length
    Ø
    Volume
    No. of openings
    Availability
    Price
  • Sampler
    Stainless steel 316L
    1500 mm
    40 mm
    4
    Log in for availability
    Log in for availability
    Each
    Each - $1,470.00
    $1,470.00
    Each
       $1,470.00
    Supplier Number: 1290K-1500
  • Sampler
    Stainless steel 316
    2000 mm
    40 mm
    4
    Log in for availability
    Log in for availability
    Each
    Each - $1,540.00
    $1,540.00
    Each
       $1,540.00
    Supplier Number: 1290K-2000
  • Sampler
    Stainless steel 316
    1000 mm
    40 mm
    3
    Log in for availability
    Log in for availability
    Each
    Each - $1,250.00
    $1,250.00
    Each
       $1,250.00
    Supplier Number: 1290K-1000
Inserts
Product Information
  • Material
    Length
    Ø
    Volume
    No. of openings
    Availability
    Price
  • 77576-210
    Insert for MultiThief
    Insert for MultiThief
    Stainless steel 316L
    5 ml
    Log in for availability
    Log in for availability
    Each
    Each - $310.00
    $310.00
    Each
       $310.00
    Supplier Number: 1290W-05
  • 77576-212
    Insert for MultiThief
    Insert for MultiThief
    Stainless steel 316L
    10 ml
    Log in for availability
    Log in for availability
    Each
    Each - $310.00
    $310.00
    Each
       $310.00
    Supplier Number: 1290W-10
  • 77576-214
    Insert for MultiThief
    Insert for MultiThief
    Stainless steel 316L
    25 ml
    Log in for availability
    Log in for availability
    Each
    Each - $310.00
    $310.00
    Each
       $310.00
    Supplier Number: 1290W-25
  • 77576-216
    Insert for MultiThief
    Insert for MultiThief
    Stainless steel 316L
    50 ml
    Log in for availability
    Log in for availability
    Each
    Each - $310.00
    $310.00
    Each
       $310.00
    Supplier Number: 1290W-50
  • 77576-218
    Insert for MultiThief
    Insert for MultiThief
    Stainless steel 316L
    60 ml
    Log in for availability
    Log in for availability
    Each
    Each - $310.00
    $310.00
    Each
       $310.00
    Supplier Number: 1290W-60