Orders LinkContactUsLinkComponent
U-D Master Sleeve Samplers
U-D Master Sleeve Samplers
U-D Master Sleeve Samplers
Certificates
U-D Master Sleeve Samplers
U-D Master Sleeve Samplers
14 Options Available Below
Product Details & Documents
The U-D sleeve sampler was developed for accurate unit dose samples (small volume).
  • Quick and easy accurate unit dose sampling
  • Different volumes can be taken using the same sampler
  • Simply change the tip for a different volume
Designed for use with free flowing powders and granules.
Product Family Options
Complete set
Product Information
  • Material
    Length
    Volume
    Availability
    Price
  • 77576-272
    Sleeve sampler set with handles, sample chambers, sampling tips and sampler case
    Sleeve sampler set with handles, sample chambers, sampling tips and sampler case
    Log in for availability
    Log in for availability
    Each
    Each - $2,300.00
    $2,300.00
    Each
       $2,300.00
    Supplier Number: 1500S-01
Handles
Product Information
  • Material
    Length
    Volume
    Availability
    Price
  • 77576-264
    T-bar handle
    T-bar handle
    600 mm
    Log in for availability
    Log in for availability
    Each
    Each - $225.00
    $225.00
    Each
       $225.00
    Supplier Number: 1500A-01
  • 77576-266
    Handle extension
    Handle extension
    600 mm
    Log in for availability
    Log in for availability
    Each
    Each - $128.00
    $128.00
    Each
       $128.00
    Supplier Number: 1500A-02
Sample chambers
Product Information
  • Material
    Length
    Volume
    Availability
    Price
  • 77576-268
    Sample chamber (type 1)
    Sample chamber (type 1)
    0.1 - 1.2 ml
    Log in for availability
    Log in for availability
    Each
    Each - $304.00
    $304.00
    Each
       $304.00
    Supplier Number: 1500A-03
  • 77576-270
    Sample chamber (type 2)
    Sample chamber (type 2)
    1.3 - 2.5 ml
    Log in for availability
    Log in for availability
    Each
    Each - $304.00
    $304.00
    Each
       $304.00
    Supplier Number: 1500A-04
Tips
Product Information
  • Material
    Length
    Volume
    Availability
    Price
  • 77576-292
    Tip for U-D sleeve sampler
    Tip for U-D sleeve sampler
    Stainless steel 316L
    1.7 ml
    Log in for availability
    Log in for availability
    Each
    Each - $145.00
    $145.00
    Each
       $145.00
    Supplier Number: 1500W-17
  • 77576-294
    Tip for U-D sleeve sampler
    Tip for U-D sleeve sampler
    Stainless steel 316L
    1.8 ml
    Log in for availability
    Log in for availability
    Each
    Each - $145.00
    $145.00
    Each
       $145.00
    Supplier Number: 1500W-18
  • 77576-296
    Tip for U-D sleeve sampler
    Tip for U-D sleeve sampler
    Stainless steel 316L
    2.2 ml
    Log in for availability
    Log in for availability
    Each
    Each - $145.00
    $145.00
    Each
       $145.00
    Supplier Number: 1500W-22
  • 77576-274
    Tip for U-D sleeve sampler
    Tip for U-D sleeve sampler
    Stainless steel 316L
    0.1 ml
    Log in for availability
    Log in for availability
    Each
    Each - $145.00
    $145.00
    Each
       $145.00
    Supplier Number: 1500W-01
  • 77576-276
    Tip for U-D sleeve sampler
    Tip for U-D sleeve sampler
    Stainless steel 316L
    0.2 ml
    Log in for availability
    Log in for availability
    Each
    Each - $145.00
    $145.00
    Each
       $145.00
    Supplier Number: 1500W-02
  • 77576-278
    Tip for U-D sleeve sampler
    Tip for U-D sleeve sampler
    Stainless steel 316L
    0.3 ml
    Log in for availability
    Log in for availability
    Each
    Each - $145.00
    $145.00
    Each
       $145.00
    Supplier Number: 1500W-03
  • 77576-280
    Tip for U-D sleeve sampler
    Tip for U-D sleeve sampler
    Stainless steel 316L
    0.4 ml
    Log in for availability
    Log in for availability
    Each
    Each - $145.00
    $145.00
    Each
       $145.00
    Supplier Number: 1500W-04
  • 77576-282
    Tip for U-D sleeve sampler
    Tip for U-D sleeve sampler
    Stainless steel 316L
    0.8 ml
    Log in for availability
    Log in for availability
    Each
    Each - $145.00
    $145.00
    Each
       $145.00
    Supplier Number: 1500W-08
  • 77576-284
    Tip for U-D sleeve sampler
    Tip for U-D sleeve sampler
    Stainless steel 316L
    1.2 ml
    Log in for availability
    Log in for availability
    Each
    Each - $145.00
    $145.00
    Each
       $145.00
    Supplier Number: 1500W-12