Orders LinkContactUsLinkComponent
Pump Sampler Sets
Pump sampler set with hand pump, stainless steel bottle adapter, PE 100 ml bottles, PE 10 m sampling tube, tube cutter, stainless steel hose weight, and case, Stainless steel 316L
  77576-408
 :  5020S-02
Pump Sampler Sets
  77576-408
Pump Sampler Sets
Pump Sampler Sets
Pump sampler set with hand pump, stainless steel bottle adapter, PE 100 ml bottles, PE 10 m sampling tube, tube cutter, stainless steel hose weight, and case, Stainless steel 316L
$1,590.00
Each
These pump samplers allow for the quick sampling of liquids.
  • Reduced contamination by using new tube and bottles for each sample
  • Only the tube and collecting bottle come into contact with the products
  • High efficiency and ability to pump water to a height of about four meters
Suitable for low to medium viscosity liquids.
  • Description:
    Pump sampler set with hand pump, stainless steel bottle adapter, PE 100 ml bottles, PE 10 m sampling tube, tube cutter, stainless steel hose weight, and case
  • Material:
    Stainless steel 316L
Accessories
  •  
    Description
  • 77576-410
    PTFE sampling tube, 5 m
    Each - $68.30