Orders LinkContactUsLinkComponent
Mini Drums
Mini Drums
Mini Drums
Certificates
Mini Drums
9 Options Available Below
Product Details & Documents
Mini drums make for ideal storage and production containers.
  • Made from high-quality 304 stainless steel
  • Ideal for both powders and liquids
  • Supplied with a drop-on lid
  • Toggle clamps and O-Ring seals are optional extras
  • Available from stock
  • Fast delivery
This range of mini drums are ideal for the storage of both powders and liquids. They come complete with a drop-on lid. Optional extras include toggle clamps and O-Ring seals, enabling the lid to be firmly secured.
Product Family Options
Product Information
  • Capacity
    Material
    Ø
    Height
    Availability
    Price
  • 1 L
    Stainless steel 304
    100 mm
    155 mm
    Log in for availability
    Log in for availability
    Each
    Each - $338.00
    $338.00
    Each
       $338.00
    Supplier Number: A434-1
    Specifications:
    More
    Specifications
    Cat. No.
    77577-960
  • 2 L
    Stainless steel 304
    130 mm
    175 mm
    Log in for availability
    Log in for availability
    Each
    Each - $368.00
    $368.00
    Each
       $368.00
    Supplier Number: A434-2
    Specifications:
    More
    Specifications
    Cat. No.
    77577-968
  • 3 L
    Stainless steel 304
    155 mm
    185 mm
    Log in for availability
    Log in for availability
    Each
    Each - $414.00
    $414.00
    Each
       $414.00
    Supplier Number: A434-3
    Specifications:
    More
    Specifications
    Cat. No.
    77577-970
  • 5 L
    Stainless steel 304
    175 mm
    240 mm
    Log in for availability
    Log in for availability
    Each
    Each - $495.00
    $495.00
    Each
       $495.00
    Supplier Number: A434-5
    Specifications:
    More
    Specifications
    Cat. No.
    77577-972
  • 6 L
    Stainless steel 304
    175 mm
    280 mm
    Log in for availability
    Log in for availability
    Each
    Each - $505.00
    $505.00
    Each
       $505.00
    Supplier Number: A434-6
    Specifications:
    More
    Specifications
    Cat. No.
    77577-974
  • 8 L
    Stainless steel 304
    200 mm
    285 mm
    Log in for availability
    Log in for availability
    Each
    Each - $581.00
    $581.00
    Each
       $581.00
    Supplier Number: A434-8
    Specifications:
    More
    Specifications
    Cat. No.
    77577-976
  • 10 L
    Stainless steel 304
    220 mm
    295 mm
    Log in for availability
    Log in for availability
    Each
    Each - $591.00
    $591.00
    Each
       $591.00
    Supplier Number: A434-10
    Specifications:
    More
    Specifications
    Cat. No.
    77577-962
  • 12.5 L
    Stainless steel 304
    237 mm
    285 mm
    Log in for availability
    Log in for availability
    Each
    Each - $670.00
    $670.00
    Each
       $670.00
    Supplier Number: A434-12
    Specifications:
    More
    Specifications
    Cat. No.
    77577-964
  • 15 L
    Stainless steel 304
    267 mm
    285 mm
    Log in for availability
    Log in for availability
    Each
    Each - $710.00
    $710.00
    Each
       $710.00
    Supplier Number: A434-15
    Specifications:
    More
    Specifications
    Cat. No.
    77577-966
Accessories
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 10 L
    Silicone seal for mini-drum, 10 L
    77578-226
    Silicone
    Log in for availability
    Each - $48.50
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 3 L
    Silicone seal for mini-drum, 3 L
    77578-240
    Silicone
    Log in for availability
    Each - $38.10
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 15 L
    Silicone seal for mini-drum, 15 L
    77578-230
    Silicone
    Log in for availability
    Each - $63.50
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for storage container, 20 L
    Silicone seal for storage container, 20 L
    77578-234
    Silicone
    Log in for availability
    Each - $63.50
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 1 L
    Silicone seal for mini-drum, 1 L
    77578-232
    Silicone
    Log in for availability
    Each - $21.70
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 12 L
    Silicone seal for mini-drum, 12 L
    77578-228
    Silicone
    Log in for availability
    Each - $53.60
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 8 L
    Silicone seal for mini-drum, 8 L
    77578-248
    Silicone
    Log in for availability
    Each - $40.80
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for storage container, 50 L
    Silicone seal for storage container, 50 L
    77578-242
    Silicone
    Log in for availability
    Each - $62.80
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 2 L
    Silicone seal for mini-drum, 2 L
    77578-238
    Silicone
    Log in for availability
    Each - $33.60
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for storage container, 25 L
    Silicone seal for storage container, 25 L
    77578-236
    Silicone
    Log in for availability
    Each - $63.50
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 5 L
    Silicone seal for mini-drum, 5 L
    77578-244
    Silicone
    Log in for availability
    Each - $39.10
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Silicone seal for mini-drum, 6 L
    Silicone seal for mini-drum, 6 L
    77578-246
    Silicone
    Log in for availability
    Each - $38.40