ContactUsLinkComponent
X-Ray and Metal Detectable Jugs, PP, Graduated
X-Ray and Metal Detectable Jugs, PP, Graduated
Catalog # 77578-220
New Product
Supplier #: M1044B-500
undefined
X-Ray and Metal Detectable Jugs, PP, Graduated
Catalog # 77578-220
Supplier Number:  M1044B-1000
New Product
Supplier #: M1044B-500
Log in for availability
Select an Option
500 ml
Select an Option
Blue
Select an Option
77578-220
About this item
X-ray and metal detectable jugs are perfect for both laboratory and production work. They are available in 2 sizes. The bright blue colour makes the jugs easy to spot if they get left in the product.
  • Robust PP construction
  • Excellent chemical resistance
  • Non sterile
  • Full batch traceability
To ensure the cleanliness of the jugs, they are manufactured and packed in a cleanroom. The robust nature of these jugs means that they can give many years of service or for more demanding applications. They can be used as a single use disposable product straight from the bag.
Ideal for use in food, pharma, chemical industries and use with powders, granules and pastes.
Certifications: These PP jugs conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.
Packaging: Supplied individually bagged.
Frequently Bought Together