Orders LinkContactUsLinkComponent
Point Sampler
Point Sampler
Point Sampler
Point Sampler
The point sampler is designed for large samples of powders.
  • Ideal for taking large volumes of powder from a single point
  • Perfect for both free-flowing and sticky powders
  • Easy to operate
  • Available from stock
  • Fast delivery
The point sampler is able to quickly take a single, large volume sample of both free-flowing or cohesive powders. The easy-to-use design means the scraper pushes the product into the sampler when the device is rotated.
Product Family Options
  • Description
    Material
    Length
    Volume
  • Point sampler
    Stainless steel 316L
    1000 mm
    100 ml
    Each
    Each - $1,850.00
    $1,850.00
    Each
       $1,850.00
     : 1640A-1000
  • Point sampler
    Stainless steel 316L
    2000 mm
    100 ml
    Each
    Each - $1,890.00
    $1,890.00
    Each
       $1,890.00
     : 1640A-2000