MiniCorer Sampler
About this item
The MiniCorer is designed for sampling cheese, wax, butter, etc.
- Available in multipe lengths
- Made from chemically resistant 316L stainless steel
- Available from stock
- Fast delivery
The MiniCorer is an ideal sampler for sampling cheese, wax, butter, and other similar substances. Available in varying lengths and made from chemically resistant 316L stainless steel, the sampler offers a convenient, quick, and easy way to take core samples.
3 Options Available Below
Product Details & Documents
The MiniCorer is designed for sampling cheese, wax, butter, etc.
- Available in multipe lengths
- Made from chemically resistant 316L stainless steel
- Available from stock
- Fast delivery
The MiniCorer is an ideal sampler for sampling cheese, wax, butter, and other similar substances. Available in varying lengths and made from chemically resistant 316L stainless steel, the sampler offers a convenient, quick, and easy way to take core samples.
Product Family Options
Product Information