Accupod Solid Samplers
About this item
The AccuPod solid sampler is ideal for unit dose sampling of powders and granules.
- Suitable for sampling blend uniformity
- Next generation in sampling technology
- Designed to minimize sampling error
The AccuPod is perfect for use in drug development and is frequently used in sampling blend uniformity. Ideal for sampling free flowing powders and granules. Sample remains in the pod until needed.
4 Options Available Below
Product Details & Documents
The AccuPod solid sampler is ideal for unit dose sampling of powders and granules.
- Suitable for sampling blend uniformity
- Next generation in sampling technology
- Designed to minimize sampling error
The AccuPod is perfect for use in drug development and is frequently used in sampling blend uniformity. Ideal for sampling free flowing powders and granules. Sample remains in the pod until needed.
Product Family Options
Product Information
- MaterialVolumeAvailabilityPrice
- 77577-320Accupod sampler, non sterileAccupod sampler, non sterileHDPE1 mlDiscontinued-Supplier Number: 8360T-10
- 77577-322Accupod sampler, sterileAccupod sampler, sterileHDPE1 mlDiscontinued-Supplier Number: 8360T-10S
- 77577-324Accupod sampler, non sterileAccupod sampler, non sterileHDPE5 mlDiscontinued-Supplier Number: 8360T-50
- 77577-326Accupod sampler, sterileAccupod sampler, sterileHDPE5 mlDiscontinued-Supplier Number: 8360T-50S
- 78117-002Accupod sampler, non sterileSupplier Number: 8360T-10Specifications:New ProductNew Product
- 78117-141Accupod sampler, sterileSupplier Number: 8360T-10SSpecifications:New ProductNew Product
- 78117-130Accupod sampler, non sterileSupplier Number: 8360T-50Specifications:New ProductNew Product
- 78117-014Accupod sampler, sterileSupplier Number: 8360T-50SSpecifications:New ProductNew Product