Orders LinkContactUsLinkComponent
Sack Samplers
Sack Samplers
Sack Samplers
Certificates
Sack Samplers
3 Options Available Below
Product Details & Documents
These sack samplers are designed to take fast powder samples through the sides of sacks and bags.
  • Suitable for sampling free flowing powders and granules
  • 316L stainless steel construction
  • Unique range of sample collection methods available
Insert the sampler through the side of the sack. Angle sampler downwards to allow the product to flow into the sample collecting container and seal the hole using an ultra adhesive label.
Ordering information: Includes a 250 ml sampling bottle.
Product Family Options
Product Information
  • Length
    Ø
    Availability
    Price
  • 77576-232
    Sack with plain ended, Master P
    Sack with plain ended, Master P
    400 mm
    25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $182.00
    $182.00
    Each
       $182.00
    Supplier Number: 1320A-01
  • 77576-234
    Sack with end cap, Master EC
    Sack with end cap, Master EC
    420 mm
    25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $292.00
    $292.00
    Each
       $292.00
    Supplier Number: 1320B-01
  • 77576-236
    Sack with bottle adapter, Master BA
    Sack with bottle adapter, Master BA
    420 mm
    25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $272.00
    $272.00
    Each
       $272.00
    Supplier Number: 1320C-01