Orders LinkContactUsLinkComponent
Sleeve Sampler
Sleeve Sampler
Sleeve Sampler
Certificates
Sleeve Sampler
6 Options Available Below
Product Details & Documents
The sleeve sampler is specially designed for large volumes at great depths.
  • Adjustable length and bright finish design
  • Extremely strong and robust extension rods
Suitable for free-flowing powders and granules.
Product Family Options
Product Information
  • Material
    Length
    Availability
    Price
  • 77576-024
    Sample chamber, 150 ml
    Sample chamber, 150 ml
    Stainless steel 316L
    1500 mm
    Log in for availability
    Log in for availability
    Each
    Each - $425.00
    $425.00
    Each
       $425.00
    Supplier Number: 1035T-150
  • 77576-022
    Sample chamber, 150 ml
    Sample chamber, 150 ml
    Stainless steel 316L
    1500 mm
    Log in for availability
    Log in for availability
    Each
    Each - $778.00
    $778.00
    Each
       $778.00
    Supplier Number: 1035P-150
  • 77576-026
    Sample chamber, 200 ml
    Sample chamber, 200 ml
    Stainless steel 316L
    200 mm
    Log in for availability
    Log in for availability
    Each
    Each - $431.00
    $431.00
    Each
       $431.00
    Supplier Number: 1035T-200
  • 77576-014
    Heavy duty handle
    Heavy duty handle
    Stainless steel 316L
    1000 mm
    Log in for availability
    Log in for availability
    Each
    Each - $302.00
    $302.00
    Each
       $302.00
    Supplier Number: 1035A-1000
  • 77576-016
    Heavy duty handle
    Heavy duty handle
    Stainless steel 316L
    1500 mm
    Log in for availability
    Log in for availability
    Each
    Each - $336.00
    $336.00
    Each
       $336.00
    Supplier Number: 1035A-1500
  • 77576-018
    Extension rod
    Extension rod
    Stainless steel 316L
    1000 mm
    Log in for availability
    Log in for availability
    Each
    Each - $292.00
    $292.00
    Each
       $292.00
    Supplier Number: 1035E-1000