Orders LinkContactUsLinkComponent
Fluid Point Sampler
Fluid Point Sampler
Fluid Point Sampler
Certificates
Fluid Point Sampler
Fluid Point Sampler
3 Options Available Below
Product Details & Documents
The fluid point sampler is designed for taking rapid samples of free-flowing liquids.
  • Ideal for sampling free-flowing liquids from most containers
  • Great for sampling pharmaceuticals, cosmetics, and foodstuffs
  • Easily cleaned
  • Available from stock
  • Fast delivery
The fluid point sampler is great for taking samples of free-flowing liquids from a range of containers, including drums. It is made from chemically resistant 316L stainless steel to provide excellent corrosion resistance.
Product Family Options
Product Information
  • Material
    Capacity
    Length
    Ø
    Availability
    Price
  • 77576-576
    Fluid Point sampler
    Fluid Point sampler
    Stainless steel 316L
    160 ml
    600 mm
    25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $711.00
    $711.00
    Each
       $711.00
    Supplier Number: 5551S-600
  • 77576-572
    Fluid Point sampler
    Fluid Point sampler
    Stainless steel 316L
    300 ml
    1000 mm
    25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $792.00
    $792.00
    Each
       $792.00
    Supplier Number: 5551S-1000
  • 77576-574
    Fluid Point sampler
    Fluid Point sampler
    Stainless steel 316L
    450 ml
    1500 mm
    25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $894.00
    $894.00
    Each
       $894.00
    Supplier Number: 5551S-1500