Orders LinkContactUsLinkComponent
SteriWare® Spatulas, Micro, Disposable
SteriWare® Spatulas, Micro, Disposable
SteriWare® Spatulas, Micro, Disposable
Certificates
SteriWare® Spatulas, Micro, Disposable
SteriWare® Spatulas, Micro, Disposable
9 Options Available Below
Product Details & Documents
PS or HDPE, sterile or non sterile. These disposable micro spatulas are ideal for sampling small volumes.
  • Moulded and packed in a cleanroom
  • Suitable for food contact
  • Available sterile (gamma irradiated)
  • Individually packed
The narrow sampling blade enables samples to be taken from very small containers such as vials.
Product Family Options
Product Information
  • Color
    Length
    Blade length
    Capacity
    Blade width
    Sterility
    Availability
    Price
  • PS
    White
    170 mm
    40 mm
    0.3 ml
    4 mm
    Sterile
    Discontinued
    Supplier Number: 8015V-170
  • PS
    White
    170 mm
    40 mm
    0.3 ml
    4 mm
    Sterile
    Discontinued
    Supplier Number: 8015V-170S
  • PS
    White
    170 mm
    40 mm
    0.5 ml
    6 mm
    Non sterile
    Discontinued
    Supplier Number: 8015N-170
  • PS
    White
    170 mm
    40 mm
    0.5 ml
    6 mm
    Sterile
    Discontinued
    Supplier Number: 8015N-170S
  • PS
    Red
    170 mm
    40 mm
    0.5 ml
    6 mm
    Sterile
    Discontinued
    Supplier Number: 8015NR-170S
  • PS
    White
    170 mm
    40 mm
    2 ml
    13 mm
    Non sterile
    Discontinued
    Supplier Number: 8015A-170
  • PS
    White
    170 mm
    40 mm
    2 ml
    13 mm
    Sterile
    Discontinued
    Supplier Number: 8015A-170S
  • HDPE
    White
    170 mm
    40 mm
    2 ml
    13 mm
    Non sterile
    Discontinued
    Supplier Number: 8015H-170S
  • HDPE
    White
    170 mm
    40 mm
    2 ml
    13 mm
    Non sterile
    Discontinued
    Supplier Number: 8015H-170
  • PS
    White
    170 mm
    40 mm
    2 ml
    13 mm
    Non sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $132.06
    $132.06
    Case of 100
       $132.06
    Supplier Number: 8015A-170
    Specifications:
    New Product
    New Product
  • PS
    White
    170 mm
    40 mm
    2 ml
    13 mm
    Sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $156.82
    $156.82
    Case of 100
       $156.82
    Supplier Number: 8015A-170S
    Specifications:
    New Product
    New Product
  • HDPE
    White
    170 mm
    40 mm
    2 ml
    13 mm
    Non sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $180.50
    $180.50
    Case of 100
       $180.50
    Supplier Number: 8015H-170
    Specifications:
    New Product
    New Product
  • HDPE
    White
    170 mm
    40 mm
    2 ml
    13 mm
    Non sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $193.46
    $193.46
    Case of 100
       $193.46
    Supplier Number: 8015H-170S
    Specifications:
    New Product
    New Product
  • PS
    White
    170 mm
    40 mm
    0.5 ml
    6 mm
    Non sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $180.50
    $180.50
    Case of 100
       $180.50
    Supplier Number: 8015N-170
    Specifications:
    New Product
    New Product
  • PS
    White
    170 mm
    40 mm
    0.5 ml
    6 mm
    Sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $211.10
    $211.10
    Case of 100
       $211.10
    Supplier Number: 8015N-170S
    Specifications:
    New Product
    New Product
  • PS
    White
    170 mm
    40 mm
    0.3 ml
    4 mm
    Sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $160.10
    $160.10
    Case of 100
       $160.10
    Supplier Number: 8015V-170S
    Specifications:
    New Product
    New Product
  • PS
    Red
    170 mm
    40 mm
    0.5 ml
    6 mm
    Sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $205.47
    $205.47
    Case of 100
       $205.47
    Supplier Number: 8015NR-170S
    Specifications:
    New Product
    New Product
  • PS
    White
    170 mm
    40 mm
    0.3 ml
    4 mm
    Sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $145.36
    $145.36
    Case of 100
       $145.36
    Supplier Number: 8015V-170
    Specifications:
    New Product