Order Entry
Puerto Rico
ContactUsLinkComponent
Scorpion Scoop Eco Powder Scoop
Scorpion Scoop Eco Powder Scoop
Catalog # 77577-868
New Product
undefined
Scorpion Scoop Eco Powder Scoop
Catalog # 77577-868
Supplier Number:  A334-1000
New Product

Specifications

  • Sterility:
    -
  • Width:
    120 mm
  • Capacity:
    1000 ml
  • Length:
    232 mm
  • Cat. No.:
    77577-868

Specifications

About this item

The Scorpion Scoop Eco is a stainless steel powder scoop that is ideal for general industrial use.

  • Light duty design
  • Spot welded with a satin finish
  • Perfect for non hygienic applications

The Scorpion Scoop Eco is a light duty version of our Scorpion PharmaScoop. The handle on top makes scooping powder easy, similar to a traditional flour scoop.

The Scorpion Scoop Eco has a spot welded construction, this means that it is not suitable for hygienic applications in the pharmaceutical and food industries. For hygienic applications please choose a scoop from our PharmaScoop range.