Orders LinkContactUsLinkComponent
Pharmalabels 'Sampled'
Pharmalabels 'Sampled'
Pharmalabels 'Sampled'
Pharmalabels 'Sampled'
Pre-printed 'Sampled' labels feature a write-on paper front.
  • Seals holes in bags
  • The pharma range have FDA ccompliant glue
Pre-printed "Sampled" labels feature a write-on paper front and a high strength, ultra adhesive. The UA range have a foil backing to provide a stronger barrier with the product. The pharma range have an FDA compliant glue. The high strength adhesive used on these labels allows them to stick on a wide range of surfaces, including very dusty areas. The working temperature range is −20 to 70 °C (−5 to 160 °F).
Product Family Options
  • Width
    Length
    Packaging
  • 95 mm
    95 mm
    500/roll
    Each
    Each - $112.00
    $112.00
    Each
       $112.00
     : 8220P-31
  • 150 mm
    150 mm
    250/roll
    Each
    Each - $132.00
    $132.00
    Each
       $132.00
     : 8220P-031