ULTRA Liquid Funnels, USP Class VI, Double-Bagged
Supplier: SAMPLING SYSTEMS LTD
New Product
Certificates
About this item
These premium sterile funnels are double bagged and are made from high quality USP class VI HDPE. A box liner is also used giving a third layer of protection.
- Double-bagged
- Produced in Class 100,000 cleanroom, packed in Class 10,000 cleanroom
- Sterilised by gamma irradiation
- Available in both 50 and 100 mm diameter
The funnels all have an ergonomic lug on the side so handling them cleanly is easy. The funnels are designed to be used straight from the bag.
This range is ideal for demanding applications such as vaccine manufacture, GMP and cleanroom manufacturing.
Certifications: USP Class VI Medical grade polymer is FDA and EC compliant, tested to USP661.
Packaging: Supplied in a lined box.
Frequently Bought Together