Order Entry
United States
ContactUsLinkComponent
X-ray and Metal Detectable Liquid Funnels
X-ray and Metal Detectable Liquid Funnels
Catalog # 77578-208
New Product
undefined
X-ray and Metal Detectable Liquid Funnels
Catalog # 77578-208
Supplier Number:  M1023B-100
New Product

Specifications

  • Description:
    Disposable funnel, blue
  • Length:
    105 mm
  • Environmentally Preferable:
  • Cat. No.:
    77578-208
  • Top I.D.:
    100 mm

Specifications

About this item

These X-ray and metal detectable liquid funnels are perfect for use in the laboratory and production. These funnels are made from a special grade of PP that can be detected. The bright blue colour also means that they can be easily seen if left in the material in line with HACCP guidelines.

  • Robust PP construction, provides excellent chemical resistance to many products
  • Full batch traceability
  • Easy to use

To ensure the cleanliness of the funnels, they are manufactured and packed in a cleanroom. They can be used as a single use disposable product straight from the bag or reused.

Certifications: These PP funnels conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.
Packaging: Supplied individually bagged.