VWR® Cleanroom Face Mask with Extensions
77322-216
- Sterility:Sterile
- Color:Green
- Size:X-Large
- Cat. No.:77322-216
VWR® Cleanroom face masks with extensions feature polypropylene and Tyvek® layers for enhanced coverage and a fog-free fit. Gamma-sterilized for safety, they are ideal for use in pharmaceutical, biotechnology, and electronics industries, ensuring high protection in critical environments.
- Ultrasonic welded
- Green polypropylene face, Tyvek® side extensions, latex-free elastic
- Gamma radiation sterilized to 10⁻⁶ SAL
- Manufactured in a controlled environment
- Made in the USA
Cleanroom face masks are constructed with a green spunbonded polypropylene outer facing, a soft polypropylene inner facing, and a 100% meltblown polypropylene media layer. The Tyvek® side extensions enhance coverage, and an aluminum nosepiece with an adhesive strip ensures a secure fit. Gamma radiation sterilized, these masks offer both safety and traceability.
: Masks are packaged 1 per bag, 500 bags per case.