Order Entry
Puerto Rico
ContactUsLinkComponent
4-Ethynyl-1,2-difluorobenzene ≥98.0% (by GC)
Flammable
Irritant
4-Ethynyl-1,2-difluorobenzene ≥98.0% (by GC)
Catalog # TCE1349-1G
Supplier:  TCI America
New Product
CAS Number:  
Flammable
Irritant
4-Ethynyl-1,2-difluorobenzene ≥98.0% (by GC)
Catalog # TCE1349-1G
Supplier:  TCI America
Supplier Number:  E1349-1G
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 g
  • Cat. No.:
    TCE1349-1G

Specifications

Specification Test Results

  • Appearance:
    Colourless to Almost Colourless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :138.12 g/mol
Flash Pt:7 °C
UN:1993
ADR:3,II
 :GHS02
 :GHS07
CAS Number:143874-13-9
Formula:C₈H₄F₂
MDL Number:MFCD08059511