Order Entry
Puerto Rico
ContactUsLinkComponent
2-Isocyanatoethyl methacrylate ≥98.0% (by GC) stabilized
Corrosive
Toxic
Health
2-Isocyanatoethyl methacrylate ≥98.0% (by GC) stabilized
Catalog # TCI1111-25G
Supplier:  TCI America
New Product
CAS Number:  
Corrosive
Toxic
Health
2-Isocyanatoethyl methacrylate ≥98.0% (by GC) stabilized
Catalog # TCI1111-25G
Supplier:  TCI America
Supplier Number:  I1111-25G
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    25 g/mol
  • Cat. No.:
    TCI1111-25G

Specifications

Specification Test Results

  • Appearance:
    Colourless to Almost Colourless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :155.153 g/mol
Boiling Pt:211 °C
Flash Pt:97 °C
ADR:6.1,I
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
EINECS:250-284-7
UN:2810
 :GHS05
 :GHS06
 :GHS08
Storage Temperature:Refrigerator
CAS Number:30674-80-7
Formula:C₇H₉NO₃
MDL Number:MFCD00080536