Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
1,3-Dioxolane ≥98.0% (by GC) stabilised

1,3-Dioxolane ≥98.0% (by GC) stabilised

Flammable
Irritant
Health
Supplier:   TCI America
1,3-Dioxolane ≥98.0% (by GC) stabilised
Supplier:  TCI America
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:646-06-0
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00003207
  • Molecular Weight:74.08 g/mol
  • Boiling Pt:74 °C
  • Flash Pt:−4 °C
  • EINECS:211-463-5
  • UN:1166
  • ADR:3,II
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Specifications for Product Family

Specification Test Results

  • Appearance:
    Colourless to Almost Colourless clear liquid
  • Purity (GC):
    min. 98.0 %
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 25 ml
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $17.00
    $17.00
    Each(1/Itm/Bx)
       $17.00

    Supplier Number: D5539-25ML
  • 500 ml
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $40.00
    $40.00
    Each(1/Itm/Bx)
       $40.00

    Supplier Number: D5539-500ML

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.