Order Entry
Puerto Rico
ContactUsLinkComponent
Dextran sulfate sodium salt, M.W. ~8000 (from dextran)
Dextran sulfate sodium salt, M.W. ~8000 (from dextran)
Catalog # TCD5143-25G
Supplier:  TCI America
New Product
CAS Number:  
Dextran sulfate sodium salt, M.W. ~8000 (from dextran)
Catalog # TCD5143-25G
Supplier:  TCI America
Supplier Number:  D5143-25G
New Product
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    25 g
  • Cat. No.:
    TCD5143-25G

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Sulfur:
    15.0 to 20.0% (calcd.on dried substance)
  • Specific rotation [a]20/D:
    +75 to +105 deg (C=5, H2O,calcd.on dried substance)
  • limiting viscosity:
    0.018 to 0.032
  • Drying loss:
    max. 10.0 %

Specification Test Results

About this item

 :
Merck Index:14,02951
CAS Number:9011-18-1
Formula:(C₆H₇Na₃O₁₄S₃)n
MDL Number:MFCD00081551