Order Entry
Puerto Rico
ContactUsLinkComponent
6-Chloro-m-anisidine ≥98.0% (by GC)
Toxic
6-Chloro-m-anisidine ≥98.0% (by GC)
Catalog # TCC3053-5G
Supplier:  TCI America
New Product
CAS Number:  
Toxic
6-Chloro-m-anisidine ≥98.0% (by GC)
Catalog # TCC3053-5G
Supplier:  TCI America
Supplier Number:  C3053-5G
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    5 g
  • Cat. No.:
    TCC3053-5G

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to lump
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :157.6 g/mol
Boiling Pt:258 °C
EINECS:219-277-6
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:2401-24-3
Formula:C₇H₈ClNO
MDL Number:MFCD00047830