HALO® PCS Phenyl-Hexyl HPLC Columns
New Product
HALO® PCS Phenyl-Hexyl is a positively charged surface chemistry designed to deliver improved peak shapes for basic compounds. Ideal for use with low ionic strength mobile phases, HALO® PCS maintains peak symmetry at higher loading capacities and provides an alternate selectivity for other C18 bonded phases.
- Excellent peak shape and increased loading capacity for basic compounds
- UHPLC and LCMS compatible
- Alternate L11 selectivity
- Built upon Fused-Core® technology for fast, efficient, and reliable separations
Alternative PCS C18 phases available in both a 90 and 160 Å pore size for small molecule and peptide analysis.
: ISO 9001:2015 certified.
: Use guard cartridges with guard holder.
: Other dimensions columns and guard cartridges are available on request.
: Supplied with a test chromatogram and batch validation certificate.
: All packaging is recyclable.
: These columns cannot be opened or repacked.
- Catalog No:
- 77574-688
- Endcapped:
- Yes
- pH Range:
- 2 - 7
- Temperature Limit:
- 60 °C (pH 2); 40 °C (pH 7)
- USP Code:
- L11
Frequently Bought Together