ContactUsLinkComponent
SDS-PAGE Sample Buffer, 2X, (2-Mercaptoethanol-free) for Electrophoresis
SDS-PAGE Sample Buffer, 2X, (2-Mercaptoethanol-free) for Electrophoresis
Catalog # TCB5834-25ML
Supplier:  TCI America
New Product
CAS Number:  
SDS-PAGE Sample Buffer, 2X, (2-Mercaptoethanol-free) for Electrophoresis
Catalog # TCB5834-25ML
Supplier:  TCI America
Supplier Number:  B5834-25ML
New Product
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    SDS-PAGE sample buffer, 2X
  • Size:
    25 ml
  • Cat. No.:
    TCB5834-25ML
Specifications
About this item
SDS-PAGE Sample Buffer is specially formulated for protein sample preparation to based on the Laemmli Method of SDS-PAGE system.
This product contains 4% SDS, 20% glycerol, 0.01% bromophenol blue and 0.125 M Tris HCl, pH approx. 6.8. Does not contain 2-Mercaptoethanol.
Caution: Please use after bringing back to normal temperature.