To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Capacity:100 ml
- Description:Rinsing solution for cultureware to prevent cell adhesion
- Environmentally Preferable:
- Cat. No.:MSPP-07010
- Application:Cancer, neuroscience, stem cell biology
- Supplier no.:7010
Specifications
About this item
Anti-Adherence Rinsing Solution is a surfactant solution for pre-treating cultureware to reduce surface tension and prevent cell adhesion. It can be used with plates, flasks, pipettes, strainers and other cultureware.
Adherence rinsing solution is required for use with the following:
- AggreWell™ plates (e.g. Catalog MSPP-34811)
- Organoid growth media, including HepatiCult™ (Catalog MSPP-06030)
- PancreaCult™ (Catalog MSPP-06040)
Species: Human, mouse, non human primate, rat and others.
Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet ( collectively, the ‘Product Restrictions’).