Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
RoboSep™ Buffer

RoboSep™ Buffer

Marketsource
RoboSep™ Buffer
Marketsource
Certificates
RoboSep™ Buffer
RoboSep™ Buffer

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

RoboSep™ buffer is recommended for EasySep™ cell separation protocols performed by RoboSep™.

Please note that one or two bottles of buffer are included with every purchase of a RoboSep™ reagent kit.

Species: Human, mouse, non human primate, rat and others.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Product Family Options

Product Information

  • Capacity
    Application
    Availability
    Price
  • MSPP-20104
    Buffer containing FBS
    Buffer containing FBS
    250 ml
    Immunology
    Log in for availability
    Log in for availability
    Each
    Each - $51.15
    $51.15
    Each
       $51.15
    Minimum order quantity is 1

    Supplier Number: 20104
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource
  • MSPP-20124
    5X concentrate
    5X concentrate
    250 ml
    Immunology
    Log in for availability
    Log in for availability
    Each
    Each - $171.00
    $171.00
    Each
       $171.00

    Supplier Number: 20124
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.