Orders LinkContactUsLinkComponent
Sample Lance
Sample Lance
Sample Lance
Certificates
Sample Lance
8 Options Available Below
Product Details & Documents
The Sample Lance is excellent for taking cross sectional samples of powders and some pastes.
  • Easy to use
  • Great for hard and dense powders
  • Available with a range of different handles
  • Made from high quality 316L stainless steel
The Sample Lance is excellent for sampling powders and pastes. Various models allow for good cross sectional sampling of powders with varying properties. If the powder is hard and dense, then the addition of the heavy-duty handle is a convenient feature.
Product Family Options
Product Information
  • Material
    Length
    Ø
    Availability
    Price
  • 77576-320
    Sample Lance, 25% opening, plain handle
    Sample Lance, 25% opening, plain handle
    Stainless steel 316L
    600 mm
    25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $171.00
    $171.00
    Each
       $171.00
    Supplier Number: 1660P-600
  • 77576-318
    Sample Lance, 25% opening, plain handle
    Sample Lance, 25% opening, plain handle
    Stainless steel 316L
    1000 mm
    25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $239.00
    $239.00
    Each
       $239.00
    Supplier Number: 1660P-1000
  • 77576-316
    Sample Lance, 25% opening, heavy-duty handle
    Sample Lance, 25% opening, heavy-duty handle
    Stainless steel 316L
    600 mm
    25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $493.00
    $493.00
    Each
       $493.00
    Supplier Number: 1660H-600
  • 77576-314
    Sample Lance, 25% opening, heavy-duty handle
    Sample Lance, 25% opening, heavy-duty handle
    Stainless steel 316L
    1000 mm
    25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $543.00
    $543.00
    Each
       $543.00
    Supplier Number: 1660H-1000
  • 77576-328
    Sample Lance, 50% opening, plain handle
    Sample Lance, 50% opening, plain handle
    Stainless steel 316L
    600 mm
    25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $173.00
    $173.00
    Each
       $173.00
    Supplier Number: 1665P-600
  • 77576-326
    Sample Lance, 50% opening, plain handle
    Sample Lance, 50% opening, plain handle
    Stainless steel 316L
    1000 mm
    25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $239.00
    $239.00
    Each
       $239.00
    Supplier Number: 1665P-1000
  • 77576-324
    Sample Lance, 50% opening, heavy-duty handle
    Sample Lance, 50% opening, heavy-duty handle
    Stainless steel 316L
    600 mm
    25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $493.00
    $493.00
    Each
       $493.00
    Supplier Number: 1665H-600
  • 77576-322
    Sample Lance, 50% opening, heavy-duty handle
    Sample Lance, 50% opening, heavy-duty handle
    Stainless steel 316L
    1000 mm
    25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $543.00
    $543.00
    Each
       $543.00
    Supplier Number: 1665H-1000