Orders LinkContactUsLinkComponent
U-D Master Samplers
U-D Master Samplers
U-D Master Samplers
Certificates
U-D Master Samplers
15 Options Available Below
Product Details & Documents
U-D Master is designed for sampling free flowing powders. By simply changing the sample insert cells, different volumes can be sampled.
  • Operation of the unit dose powder sampler is very easy
  • Push the sampler: At the required depth rotate the handle, which will take the sample
  • After removing the sampler the sample inserts can be removed for easy emptying of the sample
Single version takes one sample from each location.
Double version takes two samples from each location.
Triple version takes three samples from each location.
Product Family Options
Product Information
  • Length
    Ø
    No. of openings
    Availability
    Price
  • 77576-098
    Single style sampler
    Single style sampler
    300 mm
    25 mm
    5
    Log in for availability
    Log in for availability
    Each
    Each - $4,100.00
    $4,100.00
    Each
       $4,100.00
    Supplier Number: 1230A-300
  • 77576-100
    Single style sampler
    Single style sampler
    600 mm
    25 mm
    10
    Log in for availability
    Log in for availability
    Each
    Each - $4,890.00
    $4,890.00
    Each
       $4,890.00
    Supplier Number: 1230A-600
  • 77576-102
    Single style sampler
    Single style sampler
    900 mm
    25 mm
    6
    Log in for availability
    Log in for availability
    Each
    Each - $5,260.00
    $5,260.00
    Each
       $5,260.00
    Supplier Number: 1230A-900
  • 77576-092
    Single style sampler
    Single style sampler
    1200 mm
    25 mm
    8
    Log in for availability
    Log in for availability
    Each
    Each - $5,870.00
    $5,870.00
    Each
       $5,870.00
    Supplier Number: 1230A-1200
  • 77576-094
    Single style sampler
    Single style sampler
    1500 mm
    25 mm
    10
    Log in for availability
    Log in for availability
    Each
    Each - $6,500.00
    $6,500.00
    Each
       $6,500.00
    Supplier Number: 1230A-1500
  • 77576-096
    Single style sampler
    Single style sampler
    1800 mm
    25 mm
    12
    Log in for availability
    Log in for availability
    Each
    Each - $7,460.00
    $7,460.00
    Each
       $7,460.00
    Supplier Number: 1230A-1800
  • 77576-110
    Double style sampler
    Double style sampler
    600 mm
    25 mm
    5
    Log in for availability
    Log in for availability
    Each
    Each - $5,740.00
    $5,740.00
    Each
       $5,740.00
    Supplier Number: 1230B-600
  • 77576-112
    Double style sampler
    Double style sampler
    900 mm
    25 mm
    7
    Log in for availability
    Log in for availability
    Each
    Each - $7,460.00
    $7,460.00
    Each
       $7,460.00
    Supplier Number: 1230B-900
  • 77576-104
    Double style sampler
    Double style sampler
    1200 mm
    25 mm
    8
    Log in for availability
    Log in for availability
    Each
    Each - $8,310.00
    $8,310.00
    Each
       $8,310.00
    Supplier Number: 1230B-1200
  • 77576-106
    Double style sampler
    Double style sampler
    1500 mm
    25 mm
    10
    Log in for availability
    Log in for availability
    Each
    Each - $9,080.00
    $9,080.00
    Each
       $9,080.00
    Supplier Number: 1230B-1500
  • 77576-108
    Double style sampler
    Double style sampler
    1800 mm
    25 mm
    12
    Log in for availability
    Log in for availability
    Each
    Each - $9,920.00
    $9,920.00
    Each
       $9,920.00
    Supplier Number: 1230B-1800
  • 77576-120
    Triple style sampler
    Triple style sampler
    900 mm
    25 mm
    5
    Log in for availability
    Log in for availability
    Each
    Each - $8,650.00
    $8,650.00
    Each
       $8,650.00
    Supplier Number: 1230C-900
  • 77576-114
    Triple style sampler
    Triple style sampler
    1200 mm
    25 mm
    7
    Log in for availability
    Log in for availability
    Each
    Each - $9,920.00
    $9,920.00
    Each
       $9,920.00
    Supplier Number: 1230C-1200
  • 77576-116
    Triple style sampler
    Triple style sampler
    1500 mm
    25 mm
    8
    Log in for availability
    Log in for availability
    Each
    Each - $11,630.00
    $11,630.00
    Each
       $11,630.00
    Supplier Number: 1230C-1500
  • 77576-118
    Triple style sampler
    Triple style sampler
    1800 mm
    25 mm
    10
    Log in for availability
    Log in for availability
    Each
    Each - $12,850.00
    $12,850.00
    Each
       $12,850.00
    Supplier Number: 1230C-1800
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3.3 ml
    Sample insert, stainless steel, 3.3 ml
    77576-188
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2.6 ml
    Sample insert, stainless steel, 2.6 ml
    77576-174
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0.8 ml
    Sample insert, stainless steel, 0.8 ml
    77576-138
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2.3 ml
    Sample insert, stainless steel, 2.3 ml
    77576-168
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1.4 ml
    Sample insert, stainless steel, 1.4 ml
    77576-150
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3.1 ml
    Sample insert, stainless steel, 3.1 ml
    77576-184
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2.9 ml
    Sample insert, stainless steel, 2.9 ml
    77576-180
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1.8 ml
    Sample insert, stainless steel, 1.8 ml
    77576-158
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0.9 ml
    Sample insert, stainless steel, 0.9 ml
    77576-140
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0.6 ml
    Sample insert, stainless steel, 0.6 ml
    77576-134
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1.6 ml
    Sample insert, stainless steel, 1.6 ml
    77576-154
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1.3 ml
    Sample insert, stainless steel, 1.3 ml
    77576-148
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0.3 ml
    Sample insert, stainless steel, 0.3 ml
    77576-128
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0.1 ml
    Sample insert, stainless steel, 0.1 ml
    77576-124
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3.6 ml
    Sample insert, stainless steel, 3.6 ml
    77576-194
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2.4 ml
    Sample insert, stainless steel, 2.4 ml
    77576-170
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3.9 ml
    Sample insert, stainless steel, 3.9 ml
    77576-200
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3.5 ml
    Sample insert, stainless steel, 3.5 ml
    77576-192
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2.0 ml
    Sample insert, stainless steel, 2.0 ml
    77576-162
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2.1 ml
    Sample insert, stainless steel, 2.1 ml
    77576-164
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1.7 ml
    Sample insert, stainless steel, 1.7 ml
    77576-156
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2.8 ml
    Sample insert, stainless steel, 2.8 ml
    77576-178
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3.4 ml
    Sample insert, stainless steel, 3.4 ml
    77576-190
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2.2 ml
    Sample insert, stainless steel, 2.2 ml
    77576-166
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0.5 ml
    Sample insert, stainless steel, 0.5 ml
    77576-132
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2.5 ml
    Sample insert, stainless steel, 2.5 ml
    77576-172
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, blank
    Sample insert, stainless steel, blank
    77576-122
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2.7 ml
    Sample insert, stainless steel, 2.7 ml
    77576-176
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0.7 ml
    Sample insert, stainless steel, 0.7 ml
    77576-136
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3.2 ml
    Sample insert, stainless steel, 3.2 ml
    77576-186
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1.5 ml
    Sample insert, stainless steel, 1.5 ml
    77576-152
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1.0 ml
    Sample insert, stainless steel, 1.0 ml
    77576-142
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1.1 ml
    Sample insert, stainless steel, 1.1 ml
    77576-144
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1.2 ml
    Sample insert, stainless steel, 1.2 ml
    77576-146
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1.9 ml
    Sample insert, stainless steel, 1.9 ml
    77576-160
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3.0 ml
    Sample insert, stainless steel, 3.0 ml
    77576-182
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3.8 ml
    Sample insert, stainless steel, 3.8 ml
    77576-198
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0.4 ml
    Sample insert, stainless steel, 0.4 ml
    77576-130
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3.7 ml
    Sample insert, stainless steel, 3.7 ml
    77576-196
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 4,0 ml
    Sample insert, stainless steel, 4,0 ml
    77576-202
    Log in for availability
    Each - $128.00
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0.2 ml
    Sample insert, stainless steel, 0.2 ml
    77576-126
    Log in for availability
    Each - $128.00