PharmaSpoons Sampling Spoons
These sampling spoons are made from stainless steel. They are ideal for transferring small amounts of powder.
- Crevice-free design
- Bright polished finish
Available in 316L or 304 stainless steel.
These sampling spoons are made from stainless steel. They are ideal for transferring small amounts of powder.
- Crevice-free design
- Bright polished finish
Available in 316L or 304 stainless steel.
Product Family Options
304 Stainless steel
- DescriptionLengthCapacity
- : A504-1
Specifications
Cat. No.77578-014 - : A504-3
Specifications
Cat. No.77578-018 - : A504-2
Specifications
Cat. No.77578-016 - : A504-4
Specifications
Cat. No.77578-020 - : A504-5
Specifications
Cat. No.77578-022
316L Stainless steel
- DescriptionLengthCapacity
- : A506-600
Specifications
Cat. No.77578-026 - : A506-1000
Specifications
Cat. No.77578-024