Orders LinkContactUsLinkComponent
SteriWare® V Spatulas for Powder Sampling
SteriWare® V Spatulas for Powder Sampling
Certificates
SteriWare® V Spatulas for Powder Sampling
SteriWare® V Spatulas for Powder Sampling
12 Options Available Below
Product Details & Documents
These V spatulas are ideal for sampling food, chemical and pharmaceuticals. These sampling tool are ideal for powders, creams and pastes. To ensure cleanliness they are manufactured and then individually packed in a class 7 cleanroom (10000).
  • Robust PS construction
  • Available in narrow 10 mm width and standard 20 mm width
  • Both non sterile and sterile available
  • Blade lengths from 100 to 250 mm
Ideal for goods in quality control or just general sampling. The two widths and three lengths give ideal combination and choice.
 : Both EU and FDA compliant.
 : Individually bagged.
Product Family Options
Product Information
  • Color
    Length
    Blade width
    Sterility
    Availability
    Price
  • White
    100 mm
    20 mm
    Non sterile
    Discontinued
    Supplier Number: 8440A-100
  • White
    100 mm
    20 mm
    Sterile
    Discontinued
    Supplier Number: 8440A-100S
  • White
    150 mm
    20 mm
    Non sterile
    Discontinued
    Supplier Number: 8440A-150
  • White
    150 mm
    20 mm
    Sterile
    Discontinued
    Supplier Number: 8440A-150S
  • White
    250 mm
    20 mm
    Non sterile
    Discontinued
    Supplier Number: 8440A-250
  • White
    250 mm
    20 mm
    Sterile
    Discontinued
    Supplier Number: 8440A-250S
  • White
    100 mm
    10 mm
    Non sterile
    Discontinued
    Supplier Number: 8440N-100
  • White
    100 mm
    10 mm
    Sterile
    Discontinued
    Supplier Number: 8440N-100S
  • White
    150 mm
    10 mm
    Non sterile
    Discontinued
    Supplier Number: 8440N-150
  • White
    150 mm
    10 mm
    Sterile
    Discontinued
    Supplier Number: 8440N-150S
  • White
    250 mm
    10 mm
    Non sterile
    Discontinued
    Supplier Number: 8440N-250
  • White
    250 mm
    10 mm
    Sterile
    Discontinued
    Supplier Number: 8440N-250S
  • White
    100 mm
    10 mm
    Non sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $169.84
    $169.84
    Case of 100
       $169.84
    Supplier Number: 8440N-100
    Specifications:
    New Product
    New Product
  • White
    100 mm
    10 mm
    Sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $205.09
    $205.09
    Case of 100
       $205.09
    Supplier Number: 8440N-100S
    Specifications:
    New Product
    New Product
  • White
    150 mm
    10 mm
    Non sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $173.72
    $173.72
    Case of 100
       $173.72
    Supplier Number: 8440N-150
    Specifications:
    New Product
    New Product
  • White
    150 mm
    10 mm
    Sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $207.18
    $207.18
    Case of 100
       $207.18
    Supplier Number: 8440N-150S
    Specifications:
    New Product
    New Product
  • White
    250 mm
    10 mm
    Non sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $176.73
    $176.73
    Case of 100
       $176.73
    Supplier Number: 8440N-250
    Specifications:
    New Product
    New Product
  • White
    250 mm
    10 mm
    Sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $226.24
    $226.24
    Case of 100
       $226.24
    Supplier Number: 8440N-250S
    Specifications:
    New Product
    New Product
  • White
    100 mm
    20 mm
    Non sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $174.23
    $174.23
    Case of 100
       $174.23
    Supplier Number: 8440A-100
    Specifications:
    New Product
    New Product
  • White
    100 mm
    20 mm
    Sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $209.49
    $209.49
    Case of 100
       $209.49
    Supplier Number: 8440A-100S
    Specifications:
    New Product
    New Product
  • White
    150 mm
    20 mm
    Non sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $177.99
    $177.99
    Case of 100
       $177.99
    Supplier Number: 8440A-150
    Specifications:
    New Product
    New Product
  • White
    150 mm
    20 mm
    Sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $211.25
    $211.25
    Case of 100
       $211.25
    Supplier Number: 8440A-150S
    Specifications:
    New Product
    New Product
  • White
    250 mm
    20 mm
    Non sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $185.51
    $185.51
    Case of 100
       $185.51
    Supplier Number: 8440A-250
    Specifications:
    New Product
    New Product
  • White
    250 mm
    20 mm
    Sterile
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $235.02
    $235.02
    Case of 100
       $235.02
    Supplier Number: 8440A-250S
    Specifications:
    New Product