Order Entry
Puerto Rico
ContactUsLinkComponent
Human/Mouse Recombinant Activin A
Human/Mouse Recombinant Activin A
Catalog # MSPP-78001
CAS Number:  
undefined
Human/Mouse Recombinant Activin A
Catalog # MSPP-78001
Supplier Number:  78001
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Human cells
  • Species:
    Human
    Mouse
  • Size:
    10 µg
  • Protein/Peptide Name:
    Activin A
  • Purity:
    ≥95%
  • Cat. No.:
    MSPP-78001

Specifications

About this item

Activin A is a member of the transforming growth factor beta (TGF-β) family of proteins produced by many cell types throughout development (Gurdon et al.). It is a disulfide-linked homodimer (two beta-A chains) that binds to heteromeric complexes of a type I (Act RI-A and Act RI-B) and a type II (Act RII-A and Act RII-B) serine-threonine kinase receptor (Attisano et al.). Activins primarily signal through SMAD2/3 proteins to regulate a variety of functions, including cell proliferation, differentiation, wound healing, apoptosis, and metabolism (McDowell et al.). Activin A maintains the undifferentiated state of human embryonic stem cells (James et al.; Xiao et al.) and also facilitates differentiation of human embryonic stem cells into definitive endoderm (D’Amour et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').