Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant bFGF, ACF
Human Recombinant bFGF, ACF
Certificates
undefined
Human Recombinant bFGF, ACF

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Protein/Peptide Name:
    bFGF, ACF
  • Purity:
    ≥95%

Specifications

Frequently Bought Together